Orders LinkContactUsLinkComponent
Torque-Rite® Nuts, Rubber Fab
 :   Rubber Fab
Torque-Rite® Nuts, Rubber Fab
Torque-Rite® Nuts, Rubber Fab
Torque-Rite® allows you to control compression and expansion while maintaining constant inch/pounds force insuring a perfectsSurface I.D
when used with a Rubber Fab sanitary gasket. Torque-Rite® eliminates the problems associated with over - or under-tightening a gasket which can lead to an unsanitary system.
Product Family Options
  • Description
  • Torque-Rite® Nut, 30 in/lbs.
    Each
    Each - $647.56
    $647.56
    Each
       $647.56
     : TR-30
  • Torque-Rite® Nut, 50 in/lbs.
     : -