About this item
These BioStor™ Multi-Purpose Containers with resealable lids can be used to store almost any liquid, powder or solid
Contents are visible on contact with translucent plastic walls. Priced low enough to be disposable, yet sturdy enough for reuse. Lids may be custom imprinted for large volume users with specialized applications.
They are ideal for laboratory or hospital use. Injection molded of polypropylene or linear polyethylene, containers are virtually indestructible in normal use. Inert to most lab chemicals including formaldehyde, weak acids, organic solvents, and all bases. Stable, wide-mouth design allows for ease of filling and is less likely to tip over, even when stacked. Tight-fitting, snap-cap lids keep odors in and minimize chance of leakage