ContactUsLinkComponent
Jacketed Flask with Polished Joint, Ace Glass Incorporated
Jacketed Flask with Polished Joint, Ace Glass Incorporated
Catalog # 89074-576
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Jacketed Flask with Polished Joint, Ace Glass Incorporated
Catalog # 89074-576
Supplier:  Ace Glass
Supplier Number:  9450-08
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacity:
    100
  • Description:
    Jacketed Flask with Polished Joint
  • Cat. no.:
    89074-576
  • Supplier No.:
    9450-08
Specifications
About this item
Jacketed version of standard flasks
Furnished with polished standard taper 45/50 outer joints so that it can be used with fitting heads.