Orders LinkContactUsLinkComponent
Adjustable Jacobs Style Chuck
Adjustable Jacobs Style Chuck
Supplier:   Ace Glass
Adjustable Jacobs Style Chuck
Supplier:  Ace Glass
Certificates
Adjustable Jacobs Style Chuck
Adjustable Jacobs Style Chuck
1 Options Available Below
Product Details & Documents
Adjustable, Jacobs Style chucks to accomodate 1/16" to 3/8" diameter stirring shafts.
  • Propeller and Mixing Shafts are made of 303/304 Stainless
  • Propellers are pitched 15° to create axial flow
  • Adjustable Chucks should be ordered based upon the diameter of the motor shaft
Code -02 fits 1/4" diameter motor shafts, code -04 fits 5/16" diameter motor shafts.
Delivery information: Includes chuck key.
Product Family Options
Product Information
  • Availability
    Price
  • 89077-784
    5/16" Motor Shaft Jacobs Chuck for Stir Shafts up to 3/8" O.D.
    5/16" Motor Shaft Jacobs Chuck for Stir Shafts up to 3/8" O.D.
    Log in for availability
    Log in for availability
    Each
    Each - $188.61
    $188.61
    New Customer Promotional Price
    Each
       $188.61
    Supplier Number: 13588-04
  • Discontinued
    Supplier Number: -