Order Entry
Canada
ContactUsLinkComponent
Silicone Rubber Septum with PTFE Coating on One Side, Ace Glass Incorporated
Silicone Rubber Septum with PTFE Coating on One Side, Ace Glass Incorporated
Catalog # 89077-536
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Silicone Rubber Septum with PTFE Coating on One Side, Ace Glass Incorporated
Catalog # 89077-536
Supplier:  Ace Glass
Supplier Number:  12898-24
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Ø:
    6 mm
  • Description:
    Septum
  • Cat. no.:
    89077-536
  • Supplier No.:
    12898-24

Specifications

About this item

A thicker, 3.5 mm bonded PTFE faced silicone flat septa.