Peptide Vessel Shaker, Ace Glass Incorporated
Catalog #: 89078-082
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cat. no.:89078-082
- Electrical:120 V, 50/60
- Speed:2–30 rpm
- Supplier No.:13814-20
Specifications
About this item
This is a 180 degree wrist type action shaker that simulates shaking by hand, agitation rate can be varied from 2 to 30 rpm.
Will shake two flasks simultaneously. Two laboratory clamps hold glassware securely. Shaft bearing permits operation in a cold room or incubator. DC filtering allows the motor to run cooler and promotes longer motor life. Base configuration prevents walking. Fuse protected, with on/ off switch, variable speed control knob and six foot 3-wire grounded cord
Customers Who Bought This Also Bought