ContactUsLinkComponent
iMiniPlus PDF® Temperature Logger, Cryopak Verification Technologies
iMiniPlus PDF® Temperature Logger, Cryopak Verification Technologies
Certificates
undefined
iMiniPlus PDF® Temperature Logger, Cryopak Verification Technologies
Specifications
  • Catalog No:
  • 10033-826
  • 10033-824
  • 10033-822
  • Sensor:
  • Int. + ext. sensors
  • Int. sensor
  • Int. sensor
  • Alarm:
  • 2 alarm levels (1 high and 1 low)
  • 2 alarm levels (1 high and 1 low)
  • 2 alarm levels (1 high and 1 low)
  • Interface:
  • USB
  • USB
  • USB
  • Software:
  • Console Pro to Program and Secure Download; PDF Reader Only to View Log Trip Report
  • Console Pro to Program and Secure Download; PDF Reader Only to View Log Trip Report
  • Console Pro to Program and Secure Download; PDF Reader Only to View Log Trip Report
  • Battery life:
  • 1 - 2 years
  • 1 - 2 years
  • 1 - 2 years
  • Response time:
  • T90 of 5 min in moving air
  • T90 of 20 min in moving air
  • T90 of 20 min in moving air
  • Weight:
  • 70 g
  • 70 g
  • 70 g
  • W×D×H:
  • 83×57×18 mm
  • 83×57×18 mm
  • 83×57×18 mm
  • Data memory - No. of readings:
  • 16000
  • 16000
  • 8000
  • Batteries:
  • User replaceable
  • User replaceable
  • User replaceable