Orders LinkContactUsLinkComponent
Conductivity Standards, Sachets
Conductivity Standards, Sachets
Supplier:  Mettler Toledo
Certificates
Conductivity Standards, Sachets
Conductivity Standards, Sachets
2 Options Available Below
Product Details & Documents
NIST-traceable standard for conductivity electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Mettler Toledo sachets ensure a fresh portion of standard is used for each calibration, improving your conductivity and TDS data.
Product Family Options
Product Information
  • Size
    Conductivity
    Pack type
    Availability
    Price
  • 30×20 ml
    12.88 mS/cm
    Sachets
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302050
  • 30×20 ml
    1413 uS/cm
    Sachets
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $169.00
    $169.00
    Pack of 30
       $169.00
    Supplier Number: 51302049