KIMTECH PURE® A5 Sterile Integrated Hood and Mask with CLEAN-DON™ Technology, KIMBERLY-CLARK PROFESSIONAL®
CA89493-012
- Clothing Size:One size
- Sterile:+
- Garment application:Cleanroom
- Color:White
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA89493-012
- Supplier No.:36072
Integrated hood/mask is sonically welded together to eliminate exposure gaps
- Sonically welded to eliminate exposure gaps
- CLEAN-DON™ technology makes donning easier and reduces the risk of contamination
- Pull-down ties are located in the back to assist in the gowning process
- Stretch fit elastic and latex-free
- Type 6 PPE Category III
- Each hood/mask offers SAL 10-6
Innovative CLEAN-DON™ technology makes donning easier and reduces the risk of contamination. A blue line along the garment interior indicates the proper place to grasp while donning, helping workers avoid touching the garment exterior. Pull-down ties are located in the back to assist in the gowning process. Stretch fit elastic and latex-free. Type 6 PPE Category III for “parts of the body”. Supplied in vacuum-sealed packaging. Each hood/mask offers a sterility assurance level of 10-6.
: ISO Class 5 or higher cleanroom environments.