ContactUsLinkComponent
76 results for Crucibles
You searched for: Crucibles
Crucibles
Crucibles contain solutions to be dried for constant weight using extreme temperatures then to be ashed for minimizing weight. Often featuring a perforated bottom, the vessels serve as a filter depositing precipitates. The high grade containers are offered in both high and low form styles with or without lids. Stable crucibles are resistant to even harsh substances. Available in a variety of capacities, manufacturing, and glazing options to easily modify based on procedure and sample.
Sort by
VWR® Heavy-Duty Crucibles and Covers, Steel
VWR® Heavy-Duty Crucibles and Covers, Steel
Supplier: VWR International
Low form crucibles and covers are designed for high-temperature applications, such as alkali and peroxide fusions.
Expand 5 Items
Loading...
VWR® Bitumen Crucible, Porcelain
VWR® Bitumen Crucible, Porcelain
Supplier: VWR International
Chemical-porcelain bitumen crucible features a perforated bottom and large filtering surface.
Expand 1 Items
Loading...
VWR® Nickel Crucibles and Covers
VWR® Nickel Crucibles and Covers
Supplier: VWR International
High form crucibles and covers are drawn from one-piece, pure sheet nickel.
Expand 6 Items
Loading...
VWR® Rose Crucible, Porcelain
VWR® Rose Crucible, Porcelain
Supplier: VWR International
Unglazed rose crucible without perforated cover or tube.
Expand 1 Items
Loading...
VWR® Gooch Crucibles, Porcelain
VWR® Gooch Crucibles, Porcelain
Supplier: VWR International
Chemical-porcelain gooch crucibles are used in total suspended solids determinations.
Expand 5 Items
Loading...
Porcelain Crucibles with Covers
Porcelain Crucibles with Covers
Supplier: VWR International
These crucibles are great for student use.
Expand 5 Items
Loading...
VWR® Crucibles, PTFE
VWR® Crucibles, PTFE
Supplier: VWR International
Isostatically molded, pure FDA approved PTFE crucibles are totally chemically resistant and inert.
Expand 4 Items
Loading...
VWR® Annealing Cup, Porcelain
VWR® Annealing Cup, Porcelain
Supplier: VWR International
Chemical-porcelain annealing cup is glazed inside and out, except for outside bottom surface and rim.
Expand 1 Items
Loading...
VWR® Zirconium Crucibles
VWR® Zirconium Crucibles
Supplier: VWR International
VWR® Zirconium Crucibles are low form and are great for high-temperature fusion.
Expand 3 Items
Loading...
VWR® High and Low Form Porcelain Crucibles
VWR® High and Low Form Porcelain Crucibles
Supplier: VWR International
Chemical-porcelain crucibles are glazed inside and out, except the outside bottom and rim.
Expand 18 Items
Loading...
Continental Fire Assay Crucibles, Anachemia
Continental Fire Assay Crucibles, Anachemia
Supplier: VWR International
Continental Crucibles are specifically designed for precious metal analysis
Expand 3 Items
Loading...
VWR® Porous-Bottom Crucibles, Porcelain
VWR® Porous-Bottom Crucibles, Porcelain
Supplier: VWR International
Chemical-porcelain crucibles feature a porous bottom with a porous disc permanently formed into the walls.
Expand 3 Items
Loading...
Hunter Fire Assay Cupels, MGO, Anachemia
Hunter Fire Assay Cupels, MGO, Anachemia
Supplier: Avantor
Hunter cupels have been a consistent mainstay in the assay industry for over 30 years
Expand 1 Items
Loading...
Alpine Fire Assay Cupels, MgO, Anachemia
Alpine Fire Assay Cupels, MgO, Anachemia
Supplier: Anachemia Mining
Alpine cupels are made from high purity magnesium oxide and offer superior mechanical strength free from cracking and pitting
Expand 8 Items
Loading...
Hunter Fire Assay Cupels, Bone Ash, Anachemia
Hunter Fire Assay Cupels, Bone Ash, Anachemia
Supplier: Avantor
Hunter cupels have been a consistent mainstay in the assay industry for over 30 years
Expand 6 Items
Loading...
Straight Wall Nickel Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
In the analytical laboratory, nickel crucibles offer high resistance to dilute alkalies at a very low cost per crucible.
Expand 1 Items
Loading...
Crucibles for Hydrolysis Unit, VELP Scientifica
Expand 1 Items
Loading...
Crucibles, Tammann, Aluminum Oxide, Thermo Scientific
Supplier: Thermo Scientific Chemicals
These crucibles according to Tammann are manufactured from aluminium oxide
Expand 5 Items
Loading...
Crucibles, Cylindrical, Flat Base, Aluminum Oxide (Al-23), Thermo Scientific
Supplier: Thermo Scientific Chemicals
These crucibles are manufactured from aluminium oxide
Expand 7 Items
Loading...
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Zirconium crucibles are handmade to an exacting tolerance for uniform wall thickness
Expand 6 Items
Loading...
Fused Quartz Crucible and Lid, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Fused quartz has many desirable properties including high chemical purity, high corrosion resistance, high melting point, extreme hardness, low coefficient of thermal expansion, excellent electrical insulation qualities, and optical transmission from ultra-violet to infra-red
Expand 2 Items
Loading...
Crucibles, Conical, Flat Base, Aluminum Oxide, Thermo Scientific
Supplier: Thermo Scientific Chemicals
These crucibles are manufactured from aluminium oxide
Expand 9 Items
Loading...
A.P. Green Fire Assay Crucibles, HarbisonWalker
A.P. Green Fire Assay Crucibles, HarbisonWalker
Supplier: HD Biosciences CO., Ltd
A. P. Green Fire Assay Crucibles have several features which make them ideal for mining assays.
Expand 9 Items
Loading...
Glassy Carbon Conical Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
Expand 5 Items
Loading...
Gooch Crucible, High Form, Ace Glass Incorporated
Gooch Crucible, High Form, Ace Glass Incorporated
Supplier: Ace Glass
High Form Gooch crucible features an integral fritted disc that is 30mm in diameter
Expand 5 Items
Loading...
Glassy Carbon Cylindrical Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Glassy carbon offers high purity, corrosion resistance, thermal stability, and a structure impermeable to both gases and liquids
Expand 5 Items
Loading...
Crucibles, Cylindrical, Flat Base, Aluminum Oxide (Al-24 and Al-25), Thermo Scientific
Supplier: Thermo Scientific Chemicals
These crucibles are manufactured from aluminium oxide
Expand 7 Items
Loading...
Magnesium Oxide Crucibles, Cylindrical with Flat Base, Thermo Scientific
Supplier: Thermo Scientific Chemicals
These high performance oxide ceramics display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases.
Expand 5 Items
Loading...
Eisco Porcelain Crucible with Lid, Short Form
Eisco Porcelain Crucible with Lid, Short Form
Supplier: Wards
High thermal performance and enhanced chemical resistance.
Expand 6 Items
Loading...
High Form Porcelain Crucibles
High Form Porcelain Crucibles
Supplier: SILICO & CHEMICO PORCELAIN SE
These crucibles are similar to wide-form porcelain crucibles but taller.
Expand 6 Items
Loading...
Sort by
Recommended for You