Order Entry
Canada
ContactUsLinkComponent
N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis, Supelco®
Irritant
N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis, Supelco®
Catalog # CA1.06237.0005
Supplier:  MilliporeSigma
CAS Number:  
undefined
Irritant
N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis, Supelco®
Catalog # CA1.06237.0005
Supplier:  MilliporeSigma
Supplier Number:  1.06237.0005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, pre-packed
  • Size:
    5G
  • Cat. no.:
    CA1.06237.0005
  • Supplier No.:
    1.06237.0005

Specifications

Specification Test Results

  • Solubility:
    30 g/l (20 °C)
  • Molar Mass:
    259.18 g/mol
  • Bulk Density:
    380 kg/m3
  • pH:
    1.0 (25 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :259.18 g/mol
Merck Index:12,06496
Melting Pt:188…190 °C
EINECS:215-981-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556