

2-Propanol, Uvasol® for spectroscopy, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-89.5 °C
- Molar Mass:60.1 g/mol
- Boiling Point:82.4 °C (1013 hPa)
- Vapor Pressure:43 hPa (20 °C)
- Flash Point:12 °C
- Refractive Index:1.378
- Explosion Limit:2 - 13.4 %(V)
- Density:0.786 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:425 °C
- Evaporation number:11
- Water absorption:1000 g/kg
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Related Information
Certificates
Search by Lot NumberAYN4_Analytical_Chemistry_-_Spectroscopy VWR Spectrophotometers
Product Literature
ghs_1.00993
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together