Silica Reagent Set, High Range, Hach
CA11014-258
: Hach
:
- Chemical method:Silicomolybdate
- Description:Silica Reagent Set, High Range
- Method:Silicomolybdate
- Cat. no.:CA11014-258
- Sample Size:5 mL
- Supplier No.:2462300
- No. of Tests:100
For high range silica determination by the Silicomolybdate method
For visual test kits that require a 5 mL sample. 100 tests.
: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.