Nalgene® Amber HDPE Carboy, Thermo Scientific
Catalog #: 71490-028
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:10 L (2.6 gal.)
- Color:Amber
- Description:Amber Carboy, with Side Handles
- Cat. no.:71490-028
- Supplier No.:2256-7020
Specifications
About this item
Protect photosensitive chemicals, reagents, buffers, and standards when mixing and storing.
The Nalgene™ amber HDPE carboy is supplied with amber polypropylene closure, and meets the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.
Customers Who Bought This Also Bought