Order Entry
Canada
ContactUsLinkComponent
Nalgene® Heavy-Duty Rectangular Carboy, Thermo Scientific
Nalgene® Heavy-Duty Rectangular Carboy, Thermo Scientific
Catalog # 16115-335
undefined
Nalgene® Heavy-Duty Rectangular Carboy, Thermo Scientific
Catalog # 16115-335
Supplier Number:  2214-0050

Specifications

  • Capacity:
    20 L (5.3 gal.)
  • Description:
    High Density Polyethylene Carboy
  • Dimensions:
    323 x 234 x 399 mm (12²³⁄₃₂ x 9⁷⁄₃₂ x 15²³⁄₃₂")
  • Cat. no.:
    16115-335
  • Supplier No.:
    2214-0050

Specifications

About this item

The Nalgene® HDPE heavy-duty rectangular carboy is a space-saving option for storing and transporting reagents and other liquids.

The carboy is graduated to contain in 5 liter and 1 gallon increments.

The rugged carboy features large handles to enable easy use, even with gloved hands.

Delivery: Supplied with valve-sealing, 70 mm closure.