Order Entry
Canada
ContactUsLinkComponent
Arium® Comfort I Type 1 and 3 Combined Water Systems
Arium® Comfort I Type 1 and 3 Combined Water Systems
Supplier:  Sartorius
Certificates
undefined
Arium® Comfort I Type 1 and 3 Combined Water Systems
Supplier:  Sartorius

Specifications

  • Catalog No:
  • CA89510-628
  • CA89510-626
  • CA89510-632
  • CA89510-634
  • CA89510-640
  • CA89510-638
  • CA89510-622
  • CA89510-620
  • CA89510-646
  • CA89510-644
  • CA89510-648
  • CA89510-652
  • Production flow rate:
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • 120 L/h
  • Electrical:
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • 120V
  • W×D×H:
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • 24 × 23 × 24"
  • Weight:
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.
  • 65 lbs.

Specifications

Related Information

Frequently Bought Together