About this item
The Nitrification Control Kit is used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems. This portable and versatile kit includes a colorimeter and reagents for Free Ammonia and Monochloramine, Nitrite, and Total Chlorine tests.
Used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Versatile kit contains the colorimeter and reagents to perform tests for Free Ammonia, Monochloramine, Nitrite, Total Chlorine (low range and high range).
1 Options Available Below
Product Details & Documents
The Nitrification Control Kit is used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems. This portable and versatile kit includes a colorimeter and reagents for Free Ammonia and Monochloramine, Nitrite, and Total Chlorine tests.
Used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Versatile kit contains the colorimeter and reagents to perform tests for Free Ammonia, Monochloramine, Nitrite, Total Chlorine (low range and high range).
Documents
ghs_2882200
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 89500-778
- CA89500-778
- Platform:
- PP, S
- PP, S
Specifications
Product Family Options
Product Information
- RangeNo. of testsAvailabilityPrice
- Free ammonia, 0.02 - 0.50 mg/L NH₃-N, monochLoramine, 0.04 - 4.50 mg/L CL₂, 0 - 0.350 mg/L NO₂-N50 - 100Discontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.89500-778PlatformPP, S - Free ammonia, 0.02 - 0.50 mg/L NH₃-N, monochLoramine, 0.04 - 4.50 mg/L CL₂, 0 - 0.350 mg/L NO₂-N50 - 100Log in for availabilityEach$7,549.33Supplier Number: 2882200Specifications:
More
Specifications
Cat. No.CA89500-778PlatformPP, S