GC Collection Tube, Ace Glass Incorporated
Catalog # 89075-192
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:GC Collection Tube
- Cat. no.:89075-192
- Supplier No.:9579-06
Specifications
About this item
Microscale gas chromatography collection tube for fraction collection followed by transfer of sample to Ace Glass' conical reaction 0.1 ml vial for storage.
A standard taper 5/5 inner joint on tube allows direct connection to heated exit port of GC column via Ace Glass' #150 tsainless steel connection adapter. Inner joint facilitates transfer since 0.1 ml vial has a standard taper 5/5 outer joint.
Frequently Bought Together