Mini Glass Vials, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Multipurpose, borosilicate glass vial designed for storage and shipment of small volumes.
Coneshaped interior allows maximum retrieval of contents by syringe or micro-pipette. Ideal for iodine131, iodine125 and other isotopes.
Ordering information: Packed in convenient, reusable plastic transport or storage boxes with hinged lid and foam cells.
Customers Who Bought This Also Bought