SP Bel-Art Draft Checker™ Airflow Detection Powder, Bel-Art Products, a part of SP
Catalog # 30181-800
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Draft Checker™ Airflow Detection Powder
- Size:1.5 g
- Cat. no.:30181-800
- Supplier No.:199390000
Specifications
About this item
A quick, easy way to test the safety of fume hood exhaust before use.
- Plastic bottle easily dispenses ultra-fine silica powder
- Inert
- Non-toxic
- Non-flammable
Just a squeeze or two from the plastic bottle dispenses a cloud of ultra-fine, lightweight silica powder, which remains suspended long enough to visually check whether or not the fume hood exhaust is functioning properly.
It is also useful for detecting drafts, gas leaks, and air currents in rooms, ducts, and exhaust systems. The amorphous silica powder is inert, non-toxic, and non-flammable.
Packaging: Packaged in a 1.5 g bottle.