SP Bel-Art LabLens® Liquid, Bel-Art Products, a part of SP
13197-193
- Description:LabLens® Liquid Cleaning Solution
- Size:473 ml (1 pt.)
- Cat. no.:13197-193
- Supplier No.:248330000
Lens cleaning solution in a 473 ml (1 pt.) polyethylene bottle with spray pump.
- Polyethylene bottle construction
- Spray pump allows easy application
Bottle contains a non-silicone fluid that protects, polishes, anti-stats, and anti-fogs both glass and plastic lenses.