About this item
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications.
- Variety of membranes
- For wide range of sample preparation
Supor® membrane offers optimal support to retain chromatography resins whilst allowing smooth flow of buffers
AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications; gross fractionation and lysate clarification; and general filtration.
AcroPrep™ Advance 96-well filter plates for protein purification applications include high throughput protein purification, screening of chromatography resins, screening of chromatography conditions, protein fractionation, and antibody purification.
Mustang® membrane is able to withstand high flow rates to render fast purification of biomolecules. Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume. Outlet tip geometry minimizes sample leakage and loss during incubation steps
Specifications
- Catalog No:
- CA89135-706
- CA89135-704
- CA89135-686
- CA89135-684
- CA97065-368
- Well bottom area:
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- 0.25 cm²
- L×W×H with lid:
- 128×86×18 mm
- 128×86×18 mm
- 128×86×18 mm
- 128×86×18 mm
- L×W×H without lid:
- 128×86×14 mm
- 128×86×14 mm
- 128×86×14 mm
- 128×86×14 mm