TLG® Cylinders, To Deliver, Class A, White Scale, with Reverse Graduations and Bumper, with LOT Calibration Certificate, SATI Glassware
TLG® Brand Cylinders come with reverse metric scales
The "TD" (to deliver) appears on each cylinder and indicates the cylinder is calibrated to pour (dispense) and that the cylinder may be used repeatedly without drying.
: Designed from ASTM Specification E1272, Class A certified.
: Safety bumpers are supplied with sizes 10 mL through 1000 mL.
- Catalog No:
- CA10823-758
- CA10823-756
- CA10823-754
- CA10823-752
- CA10823-750
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
Frequently Bought Together