ContactUsLinkComponent
Iron (Ferrous) Color Disc Test Kit, Model IR-18C, Hach
Iron (Ferrous) Color Disc Test Kit, Model IR-18C, Hach
Supplier:  Hach
Certificates
undefined
Iron (Ferrous) Color Disc Test Kit, Model IR-18C, Hach
Supplier:  Hach
Specifications
  • Method:
    Color disc/1,10 Phenanthroline