ContactUsLinkComponent
BD Taxo™ Anti-Biotic Detection Discs, BD Diagnostics
BD Taxo™ Anti-Biotic Detection Discs, BD Diagnostics
Catalog # CA90002-114
Supplier:  BD
CAS Number:  
BD Taxo™ Anti-Biotic Detection Discs, BD Diagnostics
Catalog # CA90002-114
Supplier:  BD
Supplier Number:  231039
CAS Number:  
Specifications
  • Description:
    Blank Disc Vials, ¼"
  • Cat. no.:
    CA90002-114
  • Supplier No.:
    231039
Specifications
About this item
BD Taxo™ Antibiotic Detection Discs are packaged with 50 Discs per vial or cartridge
Taxo™ brand products are easy-to-use discs and strips for classical identification of microorganisms. Each disc or strip contains specific dried chemical reagents, providing a cost effective identification and differentiation tool. The use of these discs follows APHA standard methods for the examination of dairy products.