ContactUsLinkComponent
BBL™ Taxo™ ONPG Discs , BD Diagnostics
BBL™ Taxo™ ONPG Discs , BD Diagnostics
Catalog # CA-90002-220
Supplier:  BD
BBL™ Taxo™ ONPG Discs , BD Diagnostics
Catalog # CA-90002-220
Supplier:  BD
Supplier Number:  231248
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ Taxo™ ONPG Discs
  • Environmentally Preferable:
  • Cat. no.:
    CA-90002-220
  • Supplier No.:
    231248
Specifications
About this item
Taxo™ ONPG Discs are used for the detection of lactose fermenters, especially those that do not promptly ferment lactose in some routine identification media such as Triple Sugar Iron Agar (TSI Agar) or Kligler Iron Agar
These discs are 6 mm discs made from high-quality absorbent paper impregnated with approximately 100 µg of o-nitrophenyl-β-D-galactopyranoside per disc.