GK2401C Combined pH Electrode, Hach
Supplier: Hach
Certificates
About this item
The symmetry of Red Rod electrodes means that the iso pH is the same as the zero pH, which provides highly reproducible results even when temperature fluctuations occur
The reproducible results and measuring conditions are ensured by a special bayonet head. This electrode has a wide temperature range of -10 to 100°C and a stable signal. Results are fast and accurate, with a trouble-free liquid junction.
Specifications
- Catalog No:
- CA89500-932
- Junction:
- Porous Pin
- Temperature range:
- -10 to 100 °C