Model GT-1 Glutaraldehyde Test Kit, Hach
Supplier: Hach
Certificates
About this item
From oil fields to cooling towers, hospitals to food plants, poultry farms to dental offices, Hach's Glutaraldehyde Test Kit can help monitor disinfection processes
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
The test kit measures from 0.5 to 2800 mg/L (ppm) glutaraldehyde. This simple color-disc method is low-cose and accurate. Achieve fast results with this easy-to-carry test kit.
Ordering information: Kit includes three reagents, color discs, color comparator box, bottles, sprinkler caps, syringes, cylinders, marker, ampule breaker, ultra pure water, viewing tubes, instruction sheet, and carrying case.
Frequently Bought Together