ContactUsLinkComponent
SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
Catalog # 53574-100
undefined
SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
Catalog # 53574-100
Supplier Number:  F18940-0000
Specifications
  • Description:
    Pipette Tray Rack
  • Dimensions:
    286L x 216W x 29H mm (11¼ x 8½ x 1⅛")
  • Cat. no.:
    53574-100
  • Supplier No.:
    189400000
Specifications
About this item
This styrene tray is useful in lab drawers or on a benchtop for soaking, draining, or storing pipettes.
  • Styrene construction
  • Scalloped rims hold different pipette sizes
  • Compact design
Scalloped rims hold 16 small or 7 large pipettes. Center support holds 12 short pipettes.