SP Bel-Art Rack Slanter, Bel-Art Products, a part of SP
Catalog # 89093-458
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Rack Slanter
- Cat. no.:89093-458
- Supplier No.:188900100
Specifications
About this item
Tilt test tube racks to 25°, 35°, or 45°.
- Adjustable angle
- Ideal for creating agar slant tubes
- Built-in magnifier aids in viewing Petri dishes
- Autoclavable
Adjusting the angle of racks reduces hand strain when working with pipettors
Base legs have predetermined slots that the stand slides in and out of to easily change angles. Fits racks up to 29.3×24.2×8.3 cm (119/16×99/16×31/4"). Steam autoclavable at 121 °C (250 °F).
Also ideal for creating agar slant tubes or when using the Mini-Magnifier (54848-306) to aid in the viewing of Petri dishes.