Protease Inhibitor Cocktail Set I, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases
Lyophilized. Reconstitute each vial with 1 mL dH2O to yield a 100X stock solution. Water solubility.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.