You searched for: Venting Filters
For the safety of the workers and your equipment, always use a vent filter. These devices act as barriers on air lines. They contain hydrophobic media, which prevent the entry of water and aerosols into sensitive equipment and also protect the lab environment from aerosolized pathogens. Vent filters can also enable air to enter and exit vessels such as bioreactors while maintaining the sterility of the interior environment.
VWR® Gas Venting Filters
Supplier: VWR International
VWR® Gas venting filters are designed for obstructing bacterial, particles or moisture when venting in different applications. Sterile venting of bioreactors, fermentation tanks, media flasks, and carboys. Sterile gas purge of cell culture vessels or filling vessels. In-line sterilization of and particulate removal from air and gases. Obstruct moisture to protect equipment.
Expand 9 Items
Vacushield™ Vent Device, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
This disposable protective device is designed to prevent aqueous liquid and aerosol contamination of vacuum pumps or aspiration suction systems.
Expand 1 Items
Exhaust Filters V2.0, SCAT
Supplier: S.C.A.T.
The SCAT Exhaust Air Filters V2.0 made of durable HD-PE material are high-performance exhaust air filters of the latest generation for use with SafetyWasteCaps or the Universal Waste Hubs "Jan. With its specific surface area of 1200 m²/g, the multi-component granulate is the optimal filter medium for almost all solvent vapors. Thanks to the optimized particle size of the activated carbon, escaping gases or vapors from waste disposal containers are reliably cleaned and converted back into healthy breathing air.
Expand 1 Items
Nalgene® Carboy Vent Filter, Thermo Scientific
Supplier: Thermo Fisher Scientific
For venting and solvent filtration applications and for maintaining purified water stored in carboys.
Expand 1 Items
Whatman™ HEPA-VENT Venting Filters
Supplier: Cytiva
HEPA-VENT in-line disk filters are convenient, disposable, light-weight filters with high-efficiency glass fiber media for maximum particle retention.
Expand 1 Items
Whatman™ PolyVENT™ Integral Vent Filters
Supplier: Cytiva
Whatman PolyVENT disc filters are integral venting filters that work bidirectionally to prevent contamination from entering containers like fermentation vessels during incubation.
Expand 5 Items
Double HEPA Filtration Kit, Labconco®
Supplier: Labconco
Kit provides a second level of HEPA Filtration to meet United States Pharmacopeia (USP) <800> standard for redundant HEPA Filter in series.
Expand 5 Items
Whatman™ SteriVENT™ Venting Filters
Supplier: Cytiva
SteriVENT™ venting filters with an integral hydrophobic PTFE membrane and PP housing are well suited for sterile venting of vessels and tanks during draining and filling. These filters are capable of allowing clean air to pass into the vessel while preventing microbiological contamination.
Expand 3 Items
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Supplier: Cytiva
High-retention and high flow-rate glass fiber filters for research and industrial use to remove particulates from air and gases.
Expand 4 Items
Whatman™ Carbon Cap™ Carbon Filtration Capsules, Whatman products (Cytiva)
Supplier: Cytiva
Carbon cap capsule filters are filled with activated carbon and include a pleated glass fiber filter. They are suitable for adsorption of organic vapors from air, and for removing color, organics, and chlorine from water.
Expand 2 Items
Airguard® DP® Pleated Filters, MERV 8, Total Filtration
Supplier: Total Filtration Services
DP® filters are made of 100% synthetic fibers that provide mechanical efficiency for a MERV 8 performance.
Expand 4 Items
EZBio® Vent Filter, 0.22 µm, Nonsterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
EZBio® PTFE vent filters are used with EZBio® Single-Use bottle assemblies, which are perfect for fluid transfer and storage.
Expand 8 Items
ECD Vent Trap, Restek
Supplier: Restek
The ECD vent trap reduces the release of hazardous materials from the ECD vent into the lab.
Expand 1 Items
Airguard® Activator™ Self-Sealing Panel Air Filters, Total Filtration
Supplier: Total Filtration Services
Activator™ series panels and links consist of layers of multi-denier polyester media sealed around a heavy-duty 9-gauge internal wire frame with integral cross wire support members.
Expand 3 Items
Organic Vapor Filter , Quality Lab Accessories
Supplier: QLA
Organic vapor filter.
Expand 1 Items
Exhaust Filters, V3.0, S.C.A.T.
Supplier: SCAT AMERICAS, INC.
The SCAT V3.0 exhaust filter for safety waste caps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types.
Expand 8 Items
RigidAir™ Spun Glass or Synthetic Media Rigid Filter, Total Filtration
Supplier: Total Filtration Services
Rigid-Air extended media surface rigid filters are designed for use in most commercial or industrial HVAC systems where medium to high efficiency filtration is required.
Expand 1 Items
Airguard® Synthetic Pocket Filters, Total Filtration
Supplier: Total Filtration Services
Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Expand 43 Items
Airguard® Synthetic Pocket Filters, Total Filtration
Supplier: Total Filtration Services
Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Expand 29 Items
HEPA Vacuum Filter, Mortech®
Supplier: Mortech Manufacturing
This replacement filter is to be used with Mopec 1000 autopsy saw (76498-340) and/or the Mopec 810 autopsy saw (76498-414).
Expand 1 Items
Replacement Cast Vacuum Filter For Stryker 986 Cast Vac, Mortech
Supplier: Mortech Manufacturing
The BD986-500 vacuum filter is a brand new direct replacement for the Stryker 986-500 Cast Saw Vac filter. It can be used in both the Styker 986 Mobile and 987 Wall Mount vacuums. Replacing the filter every 6 months more more is recommended depending on usage.
Expand 1 Items
Filter Kit for BD093 Bone Dust Collector, Mortech®
Supplier: Mortech Manufacturing
Contains all required components for operation. Designed specifically for use with the Mopec Swordfish Vacuum Filtration System.
Expand 1 Items
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Supplier: Cytiva
Whatman HEPA-CAP Vent Filters are high-retention and high flow-rate glass fiber filters that are commonly used in both research and industry to remove particulates from air and gases.