Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art Petri Dish Incubation Tray, Bel-Art Products, a part of SP
SP Bel-Art Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Catalog # 10600-116
undefined
SP Bel-Art Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Catalog # 10600-116
Supplier Number:  F18983-0000

Specifications

  • Description:
    Petri Dish Incubation Tray
  • Dimensions:
    34.9L x 13.7W x 2.2H cm (133/4 x 53/8 x 7/8")
  • Cat. no.:
    10600-116
  • Supplier No.:
    189830000

Specifications

About this item

The petri dish incubation tray holds 100 mm (4") petri dishes securely on an incubator rack.

  • Incubation tray is an excellent aid for keeping cell cultures organized
  • Steam autoclavable at 121 °C (250 °F) for 15 to 20 minutes

Each tray has 28 holes in the bottom, 19 mm (3/4") in diameter, to allow air passage. Made from 1.6 mm (1/16") thick, white epoxy-coated steel.