ContactUsLinkComponent
Autoclavable Water Bottle, Tecniplast®
Autoclavable Water Bottle, Tecniplast®
Catalog #: 10717-475
Supplier:  Tecniplast Usa
Product available on GSA Advantage®
Autoclavable Water Bottle, Tecniplast®
Catalog #: 10717-475
Supplier:  Tecniplast Usa
Product available on GSA Advantage®
Specifications
  • Capacity:
    150 ml (5 oz.)
  • Description:
    Water Bottle, without Caps
  • Material:
    Polycarbonate
  • Cat. no.:
    10717-475
  • Supplier No.:
    ACBT0150
Specifications
About this item
Shatterproof, transparent, autoclavable water bottle with permanent graduations.
Bottles fit under filter-style covers. Designed for cage sizes A and B.
Customers Who Bought This Also Bought