BBL™ Trypticase™ Soy Broth Serum, BD Life Sciences
Catalog # CA90005-174
Supplier: BD
CAS Number:
Specifications
- Description:BBL™ Trypticase™ Soy Broth Serum
- Size:100 mL
- Cat. no.:CA90005-174
- Supplier No.:299107
Specifications
About this item
BBL™ Trypticase™ Soy Broth Serum is a prepared, bottled media for sterility, microbial limit, food, dairy, and water testing.