SP Bel-Art Wash Bottle, 2-Color, Safety-Labeled, Wide-Mouth, LDPE, Bel-Art Products, a part of SP
Bel-Art’s Safety Labeled wash bottles for dispensing your most common solvents.
- Polypropylene closure with leakproof design
- Easy to read
- LDPE solvent-resistant
Printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making the workplace a safer place.
: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
: Assortment includes one each of Acetone, Ethanol, Isopropanol, Methanol, Toluene, and Water. Use only with solvents compatible with LDPE.
Frequently Bought Together