Order Entry
Canada
ContactUsLinkComponent
ATP Solution for Ligation Reactions, Teknova
ATP Solution for Ligation Reactions, Teknova
Catalog # CA100216-332
Supplier:  Teknova
CAS Number:  
ATP Solution for Ligation Reactions, Teknova
Catalog # CA100216-332
Supplier:  Teknova
Supplier Number:  A1204
CAS Number:  

Specifications

  • Application:
  • Description:
    ATP Solution for Ligation Reactions pH 7.5
  • Size:
    300 μL
  • Environmentally Preferable:
  • Cat. no.:
    CA100216-332
  • Supplier No.:
    A1204

Specifications

About this item

Clear sterile solution in water, pH adjusted to 7.5

  • For Ligation and other Molecular Biology Applications

Tested for use in ligation reactions.