You searched for: Mortars and Pestles
Mortars and pestles are made of a variety of materials, from clear, heavy glass to hard silica minerals to porcelain and more. All components are easy to clean and provide a high degree of durability for long life despite continuous, sometimes intense use. Mortars and pestles are an enduring and ancient tool used since ancient times in sample preparation for chemical analysis procedures.
VWR® Mortars and Pestles, Agate
Supplier: VWR International
Mortars and pestles are made of thick-walled agate, which is the most common variety of chalcedony, a semiprecious, very hard silica mineral.
Expand 17 Items
VWR® Mortars and Pestles, Porcelain
Supplier: VWR International
Mortars are constructed of chemical porcelain and feature a lip and a glazed exterior.
Expand 20 Items
Mortars with Pestles, Agate, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Agate
Expand 2 Items
SP Bel-Art Liquid Nitrogen Cooled Mini Mortar and Pestle Set, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Stainless steel mortar has a welded fixture to hold a 1.5 mL microcentrifuge tube.
Expand 1 Items
SP Bel-Art Liquid Nitrogen Cooled Mortar, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Grind frozen samples without liquid nitrogen hazards.
Expand 2 Items
Mortar and Pestle Sets, Glass, Variety Glass
Supplier: Variety Glass
Mortar and pestle sets are made of clear, heavy, flint glass
Expand 5 Items
Mortars and Pestles, Sapphire, Diamonite™
Supplier: Cerco
Grinds materials not readily reduced in ceramic or metal mortars
Expand 2 Items
Mortar and Pestle Set, Agate Stone, United Scientific Supplies
Supplier: United Scientific Supplies
Mortar and Pestle Set, Agate Stone, United Scientific Supplies
Expand 6 Items
Marisco Ceramic Mortar and Pestle Sets, John M Maris Co
Supplier: John M Maris
Glazed ceramic Mortar and Pestle sets.
Expand 3 Items
Mortar and Pestle Set, Porcelain, Fully Glazed, United Scientific Supplies
Supplier: United Scientific Supplies
Mortar and Pestle Set, Porcelain, Fully Glazed, United Scientific Supplies
Expand 4 Items
Mortar and Pestle Sets
Supplier: SILICO & CHEMICO PORCELAIN SE
Oversized Pestle Provides a Comfortable Grip when Grinding
Expand 2 Items
Eisco Mortars and Pestles
Supplier: Wards
Mortar and Pestle sets made of solid porcelain with spout and unglazed grinding surfaces.
Expand 7 Items
Impact Mortar and Pestle, Chemplex®
Supplier: Chemplex
Crushes and pulverizes rocks and similar sample materials for spectrochemical analysis
Expand 1 Items
Eisco Heavy-Duty Mortars and Pestles
Supplier: Wards
Heavy-Duty Mortar and Pestle sets made of solid porcelain with thicker walls, spout, and unglazed grinding surfaces.
Expand 6 Items
Marisco Glass Mortar and Pestle Sets, John M Maris Co
Supplier: John M Maris
High quality smooth glass mortar with matching pestle.
Expand 5 Items
Mortar and Pestle Set, Porcelain, United Scientific Supplies
Supplier: United Scientific Supplies
Deep form mortar with oversized pestle.
Expand 7 Items
Porcelain Pestles
Supplier: SILICO & CHEMICO PORCELAIN SE
Porcelain Pestles
Expand 4 Items
SP Bel-Art ProCulture Extended Handle Pestle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Polypropylene tissue grinder efficiently homogenizes tissue and cells in microcentrifuge, centrifuge, and grinding tubes.
Expand 1 Items
Porcelain Mortars
Supplier: SILICO & CHEMICO PORCELAIN SE
High quality shallow mortars with pour-out lips.
Expand 4 Items
KONTES® PELLET PESTLE® Grinders, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Grinders are ideal for resuspending protein and DNA pellets or grinding soft tissues
Expand 14 Items
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
The Liquid Nitrogen-Cooled Mortar is for grinding frozen tissue samples without the hazard of liquid nitrogen droplets flying into personnel.
Expand 3 Items
SP Bel-Art Sterileware Double-Ended Pestle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Double-ended pestle has flat ends to effectively press soft tissues through screens, thereby disrupting the organ structure and producing cell isolates.
Expand 1 Items
KIMBLE® Pestles for Potter-Elvehjem Tissue Grinders, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Pestle Similar to 885500 but with a PTFE pestle and an unground tube. Sizes 21 to 24 have radial serrations on the lower portion of the pestle to deliver the homogenate into the cylindrical portion of the tube.
Expand 7 Items
Ball Pestle, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Ball Pestle is clear plexiglass.
Expand 2 Items
SP Bel-Art Disposable Polypropylene Pestles and 1.5 ml Tubes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
These disposable pestles are 8.5 cm (3½") long and are supplied with 1.5 mL microcentrifuge tubes of the same shape for optimal grinding (pestles also available without tubes).
Expand 2 Items
Monarch® Pestle Set, New England Biolabs
Supplier: New England Biolabs (NEB)
The Monarch® pestle set contains single-use microtube pestles and the accompanying 1.5 ml tubes for sample disruption and homogenization prior to nucleic acid extraction. 100 green pestles and tubes are included in this set.
Expand 1 Items
Disposable Pestles, Bio Plas
Supplier: Bio Plas
Use these polypropylene pestles to homogenize or resuspend precipitates such as protein or DNA pellets in buffer solutions
Expand 2 Items
Pestle Mixer with Matched 1.5 ml Tube, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Disposable polyethylene pestles and 1.5 mL micro centrifuge tubes are used to resuspend precipitates such as protein of DNA pellets in buffer solution, or to grind soft tissue.
Expand 2 Items
Axygen® Pre-Sterilized Individually Wrapped Blue Pestle, Corning
Supplier: Corning
Durable and inert polypropylene construction
Expand 1 Items
Hardened Steel Mortar, Retsch®
Supplier: Retsch
Designed for use with Retsch® RM200 Mortar Grinder, this hardened steel mortar is intended for use in less demanding applications, or under rough conditions