Labjaws Lab Frame Set, Ace Glass
89084-712
Product available on GSA Advantage®
Small Business Enterprise
- Description:Labjaws Lab Frame Set, Aluminum, 61 x 61cm
- Cat. no.:89084-712
- Supplier No.:11160-04
This Lab Frame Set can be assembled into a support 61cm wide and 61 cm high with a 46 cm base
Frame consists of (8) 5 cm rods, (2) 46 cm rods, (8) 61 cm rods, (18) S connectors, (4) rod end connectors and (4) feet.
It is convenient for small laboratory setups and provides a strong support to which glassware, heaters, may be attached with Castaloy clamps.