Order Entry
Canada
ContactUsLinkComponent
 
Labjaws Round Support Plate, Ace Glass
  89084-720
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Labjaws Round Support Plate, Ace Glass
  89084-720
 :  Ace Glass
 :  11172-17
Product available on GSA Advantage®
Small Business Enterprise

 

  • Ø:
    152 mm (6")
  • Description:
    Labjaws Round Support Plate, Aluminum
  • Cat. no.:
    89084-720
  • Supplier No.:
    11172-17

 

 

This Labjaws aluminum plate supports apparatus with flat base on Flexaframe lattice

Support is 15 cm (6") in diameter.