Reaction Head, 4 Neck Conical Flange, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Standard 4" (100mm) head for use with ACE reaction flasks
Supplied with three outer joints in line, plus a standard taper 10/30 joint for a ground joint thermometer or a thermometer adapter. Use ACE clamps for securing to flasks. Fabricated from borosilicate glass.
Frequently Bought Together