Order Entry
Canada
ContactUsLinkComponent
Axygen® Mag Plant Genomic DNA Kit, Corning
Axygen® Mag Plant Genomic DNA Kit, Corning
Supplier:  Corning
Certificates
undefined
Axygen® Mag Plant Genomic DNA Kit, Corning
Supplier:  Corning
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    Mag Plant Genomic DNA Kit

Specifications

Related Information

Certificates
Search by Lot Number
Frequently Bought Together