Human DBX1 Peptide (15 aa near C-terminus)
Catalog # 89421-074
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:15 aa near C-terminus
- Protein/peptide name:DBX1 Peptide (15 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89421-074
- Supplier No.:6455P
Specifications
About this item
DBX1 peptide is used for blocking the activity of DBX1 antibody.