Human AXIN2 Peptide (20 aa near C-terminus)
Catalogue # 89420-886
Fournisseur: Prosci
Numéro CAS:
Spécifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:20 aa near C-terminus
- Protein/peptide name:AXIN2 Peptide (20 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-886
- Supplier No.:6163P
Spécifications
À propos de cet article
AXIN2 peptide is used for blocking the activity of AXIN2 antibody.