Human LMBRD1 Peptide (18 aa near C-terminus)
89420-806
: Prosci
:
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:LMBRD1 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-806
- Supplier No.:6047P
LMBRD1 peptide is used for blocking the activity of LMBRD1 antibody.