Human RNF8 Peptide (14 aa near C-terminus)
89420-608
: Prosci
:
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near C-terminus
- Protein/peptide name:RNF8 Peptide (14 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-608
- Supplier No.:5763P
RNF8 peptide is used for blocking the activity of RNF8 antibody.