Human APC1 Peptide (17 aa near C-terminus)
Catalog # 89420-566
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near C-terminus
- Protein/peptide name:APC1 Peptide (17 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-566
- Supplier No.:5717P
Specifications
About this item
APC1 peptide is used for blocking the activity of APC1 antibody.