Palladium standard solution, 10,000 mg/L Pd in 10% nitric acid standard for ICP
Catalog # 89420-332
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa in the center
- Protein/peptide name:of GOLPH2 Peptide (18 aa in the center)
- Tested applications:Blocking
- Cat. no.:89420-332
- Supplier No.:5441P
Specifications
About this item
GOLPH2 peptide is used for blocking the activity of GOLPH2 antibody.