Human MATN1 Peptide (13 aa near C-terminus)
Catalog # 89420-112
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa near C-terminus
- Protein/peptide name:MATN1 Peptide (13 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-112
- Supplier No.:5125P
Specifications
About this item
MATN1 peptide is used for blocking the activity of MATN1 antibody.