Human LRFN1 Peptide (18 aa near C-terminus)
Catalog # 89420-064
Supplier: Prosci
CAS Number:
Specifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:18 aa near C-terminus
- Protein/peptide name:LRFN1 Peptide (18 aa near C-terminus)
- Tested applications:Blocking
- Cat. no.:89420-064
- Supplier No.:5065P
Specifications
About this item
LRFN1 peptide is used for blocking the activity of LRFN1 antibody.